Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix H. capsulatum-S.cerevisiae Recombinant, Titered
SDP

Catalog No. 22156849 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-849 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-849 Supplier ZeptoMetrix Supplier No. 0801676
Add to Cart
Add to Cart

Can be used to determine limit of detection (LOD), in diagnostic assay development or cross-reactivity studies

  • Each frozen aliquot contains 1mL of pure, titered culture of recombinant strain of Saccharomyces cerevisiae
  • Identification of the organism was confirmed by rDNA sequencing
  • Biosafety level 1
  • Must be used within the confines of Biological Safety cabinet

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Specifications

Biosafety Level 1
Certifications/Compliance ISO, 13485
Content And Storage -65°C
Form Solution
Media Type Vial
Product Type Titered Live Bacteria Control
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.