Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix H. capsulatum-S. cerevisiae Recombinant W303-Hca, Genomic DNA
SDP

Catalog No. 22156703 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-703 1 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Use with LUMINEX Assay only.
Catalog No. 22-156-703 Supplier ZeptoMetrix Supplier No. 0801676DNA1UG
Add to Cart
Add to Cart
Use with LUMINEX Assay only.

Used as an amplification and/or detection control for nucleic acid testing of H. capsulatum

  • Each aliquot contains 1μg of DNA extracted from pure culture of recombinant strain of Saccharomyces cerevisiae
  • Supplied in TE Buffer
  • Can also be used to determine a limit of detection (LOD), in diagnostic assay development, cross-reactivity studies or genomic sequencing

Specifications

Certifications/Compliance ISO, 13485
Content And Storage -20°C
Form Solution
Product Type Genomic DNA Bacteria Control
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.