Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC i-Inositol-d1 | 99.5% | 1 MG
SDP

Catalog No. 5000412682
Encompass_Preferred

i-Inositol-d1 is a stable isotope of inositol, with a molecular formula of C6H11DO6 and a molecular weight of 181.16. It appears as a white to off-white solid.

  • High purity (HPLC): 99.46%
  • High isotopic enrichment: 98%
  • Suitable for research use only
  • Stable in powder form for up to 3 years at -20°C
  • Stable in solvent for up to 6 months at -80°C

Catalog No. 50-004-12682 Supplier Medchemexpress LLC Supplier No. HYB1411S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

i-Inositol-d1 is a stable isotope of inositol, with a molecular formula of C6H11DO6 and a molecular weight of 181.16. It appears as a white to off-white solid.

  • High purity (HPLC): 99.46%
  • High isotopic enrichment: 98%
  • Suitable for research use only
  • Stable in powder form for up to 3 years at -20°C
  • Stable in solvent for up to 6 months at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.