Learn More
Medchemexpress LLC I-Inositol-d6 (myo-Inositol-d6) | 68922-44-1 | 99.9% | 5 MG
SDP
Catalog No. 5000413333
Encompass_Preferred
i-Inositol-d6 is a deuterium-labeled version of i-Inositol, a chemical compound whose associated lipids are commonly found in many fruits, particularly cantaloupe and oranges.
  • This compound can be used as a tracer.
  • It can also serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13333 Supplier Medchemexpress LLC Supplier No. HYB1411S5MG
i-Inositol-d6 is a deuterium-labeled version of i-Inositol, a chemical compound whose associated lipids are commonly found in many fruits, particularly cantaloupe and oranges.
  • This compound can be used as a tracer.
  • It can also serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.