Learn More
Medchemexpress LLC i-Inositol (Standard) | 87-89-8 | 99.9% | 250 MG
SDP
Catalog No. 5000414123
Encompass_Preferred
i-Inositol (Standard) is the analytical standard of i-Inositol, intended for research and analytical applications. i-Inositol (myo-Inositol) is a compound of sugar alcohols involved in biological processes such as insulin signal transduction and cytoskeletal transduction. It mainly exists in glial cells and plays an osmotic role.
  • Used as an analytical standard and reference standard assay.
  • Suitable for qualitative, quantitative, and methodological research.
  • Applicable in HPLC, GC, and MS experiments.
Catalog No. 50-004-14123 Supplier Medchemexpress LLC Supplier No. 250MGHYB1411R
i-Inositol (Standard) is the analytical standard of i-Inositol, intended for research and analytical applications. i-Inositol (myo-Inositol) is a compound of sugar alcohols involved in biological processes such as insulin signal transduction and cytoskeletal transduction. It mainly exists in glial cells and plays an osmotic role.
  • Used as an analytical standard and reference standard assay.
  • Suitable for qualitative, quantitative, and methodological research.
  • Applicable in HPLC, GC, and MS experiments.