Learn More
Cell Signaling Technology IkappaB Alpha (L27H11) Mouse Monoclonal Antibody, Unconjugated, BSA and Azide Free, 100 μg
SDP
Catalog No. 503132849
Encompass
The IkappaB alpha (L27H11) Mouse Monoclonal Antibody (BSA and Azide Free) is designed to detect endogenous levels of total IκBα protein. Produced by immunizing animals with a fragment corresponding to the amino terminus of human IκBα, this monoclonal antibody is provided in a carrier-free formulation ideal for technologies requiring specialized or custom labeling, such as fluorophores, metals, lanthanides, and oligonucleotides.
  • Validated for use in ELISA applications
  • Formulated in 1X PBS without BSA or azide
  • Quality control tested by size exclusion chromatography to ensure antibody integrity
  • Optimized for human and mouse reactivity
  • Ideal for custom conjugation workflows
Catalog No. 50-313-2849 Supplier Cell Signaling Technology Supplier No. 90155SF
The IkappaB alpha (L27H11) Mouse Monoclonal Antibody (BSA and Azide Free) is designed to detect endogenous levels of total IκBα protein. Produced by immunizing animals with a fragment corresponding to the amino terminus of human IκBα, this monoclonal antibody is provided in a carrier-free formulation ideal for technologies requiring specialized or custom labeling, such as fluorophores, metals, lanthanides, and oligonucleotides.
  • Validated for use in ELISA applications
  • Formulated in 1X PBS without BSA or azide
  • Quality control tested by size exclusion chromatography to ensure antibody integrity
  • Optimized for human and mouse reactivity
  • Ideal for custom conjugation workflows