Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Lactobacillus jensenii Z264, Titered
SDP

Catalog No. 22158056 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-056 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order
Catalog No. 22-158-056 Supplier ZeptoMetrix Supplier No. 0804260
Add to Cart
Add to Cart
Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order

Used to determine limit of detection (LOD), in diagnostic assay development. ZeptoMetrix Lactobacillus jensenii Z264, Titered is used for research, product development or quality assurance.

  • Identification of the organism was confirmed by 16S sequencing
  • Purity of the culture was monitored by gram staining and by additional culturing

Specifications

Biosafety Level 1
Certifications/Compliance ISO 13485
Content And Storage -65°C or below
Form Solution
Product Type Titered Live Bacteria Control
For Use With (Application) Limit of detection (LOD), in diagnostic assay development or cross-reactivity studies
Quantity 1 mL
Species Lactobacillus jensenii

For research use only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.