Learn More
Takara Bio iMatrix-511, 2 x 175 ug
SDP
Catalog No. NC2001958
Encompass
Propagation of embryonic stem (ES) cells or induced pluripotent stem (iPS) cells requires a culture substrate that retains pluripotency, a stable karyotype, and expression of pluripotency markers. iMatrix-511 is a recombinant laminin-511 E8 fragment product that can be used as an ES or iPS cell-culture substrate.
Catalog No. NC2001958 Supplier Takara Bio Supplier No. T303
Propagation of embryonic stem (ES) cells or induced pluripotent stem (iPS) cells requires a culture substrate that retains pluripotency, a stable karyotype, and expression of pluripotency markers. iMatrix-511 is a recombinant laminin-511 E8 fragment product that can be used as an ES or iPS cell-culture substrate.