Learn More
Reprocell iMatrix-511 Stem Cell Culture Substrate, 175 μg per tube, 6 tubes, Liquid, >95% pure, 500 μg/mL concentration, 150 kDa molecular weight, 2 years stability, Sterile, Xeno-free, Supports single-cell passage, Compatible with feeder-free culture
Catalog No. NC1283861
Encompass
iMatrix-511 is a highly purified, xeno-free recombinant laminin-511 E8 fragment designed for robust stem cell culture. This substrate, produced in CHO cells, promotes superior adhesion and proliferation of pluripotent stem cells, facilitating feeder-free culture and single-cell passage. Its liquid format ensures ease of use while maintaining potent integrin binding capacity.
  • Highly purified, xeno-free format
  • Produced in CHO cells
  • Liquid format for easy handling
  • Supports adhesion and proliferation of stem cells
  • Compatible with feeder-free culture and single-cell passage
  • Integrin binding specificity with a Kd < 10 nM
  • Stable for 2 years from manufacturing date
  • Sterile and easy to use
  • Contains 500 μg/mL concentration
  • Pack size includes 6 tubes, each with 175 μg
Catalog No. NC1283861 Supplier Reprocell Supplier No. NP892012
iMatrix-511 is a highly purified, xeno-free recombinant laminin-511 E8 fragment designed for robust stem cell culture. This substrate, produced in CHO cells, promotes superior adhesion and proliferation of pluripotent stem cells, facilitating feeder-free culture and single-cell passage. Its liquid format ensures ease of use while maintaining potent integrin binding capacity.
  • Highly purified, xeno-free format
  • Produced in CHO cells
  • Liquid format for easy handling
  • Supports adhesion and proliferation of stem cells
  • Compatible with feeder-free culture and single-cell passage
  • Integrin binding specificity with a Kd < 10 nM
  • Stable for 2 years from manufacturing date
  • Sterile and easy to use
  • Contains 500 μg/mL concentration
  • Pack size includes 6 tubes, each with 175 μg