Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, 95.0%, MilliporeSigma™

Catalog No. 50947830 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-830 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-830 Supplier MilliporeSigma Supplier No. HX0291P1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-54-3
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane

Specifications

Melting Point −94.3°C
Quantity 4 L
Flash Point −22°C
Packaging Glass Bottle (Pre-packed)
Assay Percent Range Assay (n-hexane): 95% min; Assay (total hexane isomers and methylcyclopentane): 99.0% min
Solubility Information 0.0095 g/L
Boiling Point 69°C (1013 hPa)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.