Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters I/O Connector Plug, 12 pin

Catalog No. 50793810 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The I/O Connector Plug is a removable connector found on the back of the instrument that holds the 12 pins for the input/output signals.

  • Removable connector
  • Holds 12 pins for input/output signals
  • Suitable for use as spare parts kits
  • Units per package: 1
  • Replacement schedule: As required

Catalog No. 50-793-810 Supplier Waters Corp Supplier No. WAT270868
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The I/O Connector Plug is a removable connector found on the back of the instrument that holds the 12 pins for the input/output signals.

  • Removable connector
  • Holds 12 pins for input/output signals
  • Suitable for use as spare parts kits
  • Units per package: 1
  • Replacement schedule: As required

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.