Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylene, 98.0%, MilliporeSigma™

Catalog No. 50947577 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-577 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-577 Supplier MilliporeSigma Supplier No. XX00201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS specifications for routine chemical synthesis, purification, and quality control applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point −25.2°C
Quantity 4 L
Flash Point 30°C
Packaging Glass Bottle (Pre-packed)
Assay Percent Range 98.0% min
Vapor Pressure 7 hPa (20°C)
Solubility Information 0.18 g/L
Percent Purity ≥98.0%
Grade Not Available
Chemical Name or Material o-Xylene
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.