Learn More
Cayman Chemical I-OMe AG-538, Purity: ≥95%, CAS: 1094048-77-7, 10 mg
SDP
Catalog No. 502910489
Encompass_Preferred
An IGF-1R and PI5P4Ka inhibitor (IC50s = 3.4 and 1 µM, respectively); selectively cytotoxic to nutrient-deprived PANC-1 cells over nutrient-sufficient PANC-1 cells
Catalog No. 50-291-0489 Supplier Cayman Chemical Supplier No. 3509010 MG
An IGF-1R and PI5P4Ka inhibitor (IC50s = 3.4 and 1 µM, respectively); selectively cytotoxic to nutrient-deprived PANC-1 cells over nutrient-sufficient PANC-1 cells