Learn More
Cayman Chemical I-OMe AG-538, Purity: ≥95%, CAS: 1094048-77-7, 5 mg
SDP
Catalog No. 502912682
Encompass_Preferred
An IGF-1R and PI5P4Ka inhibitor (IC50s = 3.4 and 1 µM, respectively); selectively cytotoxic to nutrient-deprived PANC-1 cells over nutrient-sufficient PANC-1 cells
Catalog No. 50-291-2682 Supplier Cayman Chemical Supplier No. 350905 MG
An IGF-1R and PI5P4Ka inhibitor (IC50s = 3.4 and 1 µM, respectively); selectively cytotoxic to nutrient-deprived PANC-1 cells over nutrient-sufficient PANC-1 cells