Learn More
Medchemexpress LLC I-OMe-tyrphostin AG 538 | 1094048-77-7 | MFCD03453582 | 98.0% | 437.19 g/mol | C17H12INO5 | 5 MG
SDP
Catalog No. 5000222149
Encompass_Preferred
I-OMe-Tyrphostin AG 538 is a small-molecule inhibitor of insulin-like growth factor 1 receptor (IGF-1R) and PI5P4Kα, provided as a yellow-to-orange solid for research use. It is used to probe IGF-1R-mediated signaling and kinase pathways in biochemical and cell-based assays; follow supplier storage recommendations for powder and solutions to maintain stability.
  • Selective IGF-1R and PI5P4Kα inhibition for pathway studies.
  • High purity (~98.0%) supports reproducible experimental results.
  • Yellow-to-orange solid formulation suitable for storage and handling.
  • Validated for biochemical and cell-based assay applications.
  • Supplied in milligram-scale quantities for research use.
Catalog No. 50-002-22149 Supplier Medchemexpress LLC Supplier No. HY1356805MG
I-OMe-Tyrphostin AG 538 is a small-molecule inhibitor of insulin-like growth factor 1 receptor (IGF-1R) and PI5P4Kα, provided as a yellow-to-orange solid for research use. It is used to probe IGF-1R-mediated signaling and kinase pathways in biochemical and cell-based assays; follow supplier storage recommendations for powder and solutions to maintain stability.
  • Selective IGF-1R and PI5P4Kα inhibition for pathway studies.
  • High purity (~98.0%) supports reproducible experimental results.
  • Yellow-to-orange solid formulation suitable for storage and handling.
  • Validated for biochemical and cell-based assay applications.
  • Supplied in milligram-scale quantities for research use.