Learn More
TARGETMOL CHEMICALS INC I-OME-TYRPHOSTIN AG 538 5MG
SDP
Catalog No. 502577450
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. I-OMe-Tyrphostin AG 538 is a specific IGF-1R inhibitor and ATP-competitive inhibitor of PI5P4K(alpha) (IC50 1 µM).I-OMe-Tyrphostin AG 538 inhibits IGF-1R-mediated signaling and exhibits preferential cytotoxicity to nutrient-deficient PANC1 cells. purity: 96%
Catalog No. 50-257-7450 Supplier TARGETMOL CHEMICALS INC Supplier No. T115935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. I-OMe-Tyrphostin AG 538 is a specific IGF-1R inhibitor and ATP-competitive inhibitor of PI5P4K(alpha) (IC50 1 µM).I-OMe-Tyrphostin AG 538 inhibits IGF-1R-mediated signaling and exhibits preferential cytotoxicity to nutrient-deficient PANC1 cells. purity: 96%