Learn More
AVANTOR MASTERFLEX (VWR LLC) C-Flex spooled tubing, I/P 73, 100 ft per spool.
Catalog No. NC0237021
Encompass
Masterflex C-Flex spooled tubing, I/P 73, 100 ft per spool.Long tubing lifeUltra smooth inner surfaceFewer extractablesSimilar to silicone with chemical compatibility of TygonNontoxic; no leachable plasticizers
Catalog No. NC0237021 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0642773/DEL
Masterflex C-Flex spooled tubing, I/P 73, 100 ft per spool.Long tubing lifeUltra smooth inner surfaceFewer extractablesSimilar to silicone with chemical compatibility of TygonNontoxic; no leachable plasticizers