Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, PharMed BPT, I/P 73; 150 ft
Catalog No. NC9348907
Encompass
Masterflex PharMed BPT Precision pump tubing is designed to withstand over 10,000 hours of tubing life, reducing downtime. Ideal for protecting sensitive fluids in biopharmaceutical applications and cell and tissue culture work, this tubing complies with USP class VI and FDA standards. The opaque tan color is good for light-sensitive samples. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide, making it suitable for clean-in-place and steam-in-place cleaning and sterilization systems. Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.
Catalog No. NC9348907 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9568773/DEL
Masterflex PharMed BPT Precision pump tubing is designed to withstand over 10,000 hours of tubing life, reducing downtime. Ideal for protecting sensitive fluids in biopharmaceutical applications and cell and tissue culture work, this tubing complies with USP class VI and FDA standards. The opaque tan color is good for light-sensitive samples. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide, making it suitable for clean-in-place and steam-in-place cleaning and sterilization systems. Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.