Learn More
Medchemexpress LLC I-PEG5-OH 100mg
SDP
Catalog No. 5000205986
Encompass_Preferred
I-PEG5-OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05986 Supplier Medchemexpress LLC Supplier No. HYW096157100MG
I-PEG5-OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]