Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Propionibacterium acnes Z144, Titered
SDP

Catalog No. 22157932 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-932 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-932 Supplier ZeptoMetrix Supplier No. 0801913
Add to Cart
Add to Cart

Live, titered microorganisms can be used to determine a limit of detection (LOD) in diagnostic assay development or cross-reactivity studies

  • Identification of organism was confirmed by 16S sequencing
  • Purity of the culture was monitored by Gram staining and by additional culturing
  • Freezing medium contains 15% glycerol as a cryoprotectant

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Content And Storage -65°C or below
Form Solution
Product Type Titered Live Bacteria Control
For Use With (Application) Determine LOD in diagnostic assay development/cross-reactivity studies
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.