Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Proteus vulgaris Z129, Titered
SDP

Catalog No. 22157825 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-825 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains live organism and is potentially biohazardous.
Catalog No. 22-157-825 Supplier ZeptoMetrix Supplier No. 0801898
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains live organism and is potentially biohazardous.

Proteus vulgaris is a gram-negative, rod-shaped bacterium exsisting in the intestinal tracts of humans and animals

  • Biosafety level 2 microorganism
  • Available in 1mL frozen aliquot
  • 16S sequencing used to confirm identification of organism
  • Gram staining with additional culturing monitors purity of culture
  • Glycerol (15%) used as cryoprotectant for freezing medium
  • Also used as control for nucleic acid tests

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Content And Storage -65°C
Product Type Titered Live Bacteria Control
For Use With (Application) In determination of limit of detection, diagnostic assay development or cross-reactivity studies and as controls in nucleic acid tests
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.