Learn More
TECHLAB™ QUIK CHEK™ Tray
Catalog No. 23900562
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
23-900-562 Plastic
1 options
This item is not returnable. View return policy
Catalog No. 23-900-562 Supplier TECHLAB™ Supplier No. T9000
Add to Cart
Add to Cart
This item is not returnable. View return policy
This QUIK CHEK™ Tray is a formed plastic tray designed to hold up to 32 QUIK CHEK™ Tray devices and all reagents necessary to perform the corresponding test.
  • May be cleaned with bleach or ethanol
Specifications
Color Natural
Material Plastic
Type Utility Tray
Autoclavable Not Autoclavable