Learn More
New England Biolabs, Inc. I-SceI – 2500 units
SDP
Catalog No. 50812399
Encompass
A restriction endonuclease that recognizes the sequence TAGGG_ATAA^CAGGGTAAT. I-SceI is an intron-encoded endonuclease. The intron encoding I-SceI is present in the mitochondria of Saccharomyces cerevisiae.
Catalog No. 50-812-399 Supplier New England Biolabs, Inc. Supplier No. R0694L
A restriction endonuclease that recognizes the sequence TAGGG_ATAA^CAGGGTAAT. I-SceI is an intron-encoded endonuclease. The intron encoding I-SceI is present in the mitochondria of Saccharomyces cerevisiae.