Learn More
Description
- Can also be used to determine a limit of detection (LOD), in diagnostic assay development, cross-reactivity studies or genomic sequencing
- Each aliquot contains 10μg of DNA extracted from pure culture
- Supplied in TE Buffer
Amplification/detection control for nucleic acid testing of Shigella sonnei
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | -20°C |
| Form | Solution |
| Product Type | Genomic DNA Bacteria Control |
| For Use With (Application) | Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
