Learn More
Invitrogen™ Silencer™ siRNA Transfection II Kit
Description
This Ambion™ kit is for the optimization of transfection protocols for specific cell types. It includes 0.4 mL of each of two transfection agents; siPORT™ Amine Transfection Agent and siPORT™ NeoFX™ Transfection Agent along with control siRNAs.
- Broad cell line compatibility-kit includes siPORT™ Amine and siPORT™ NeoFX™ transfection agents
- Faster protocol optimization-kit includes ready-to-transfect GAPDH and negative siRNA controls
The choice of transfection agent is critical for gene silencing experiments. Every transfection agent has a different delivery efficiency when used with different cell types, and the overall transfection efficiency and degree of gene silencing depend on the nature of the transfection agent/siRNA complex. Whether you are new to RNAi research or you want to expand your experiments to include different cell lines, the Silencer™ siRNA Transfection II Kit enables you to optimize your transfection protocols for specific cell types. The Silencer™ siRNA Transfection II Kit includes siPORT™ Amine Transfection Agent (a polyamine mixture) and the new siPORT™ NeoFX™ Transfection Agent (a cationic and neutral lipid mixture). These reagents support efficient, reproducible siRNA transfections in different cell types.
Furthermore, both can be used to transfect cells during subculturing-saving a full day of valuable time-without increased cytotoxicity. In addition to containing a choice of transfection agents, this kit includes a well-characterized GAPDH synthetic siRNA that can be used to optimize transfection conditions. Results can also be verified at the protein level with GAPDH-specific antibodies. A scrambled, negative control siRNA is also provided to confirm the specificity of the siRNA of interest and to demonstrate the absence of nonspecific, toxic effects.
Specifications
Specifications
| Content And Storage | siPORT™ NeoFX™ Transfection Agent and siPORT™ Amine Transfection Agent should be stored at 4°C. GAPDH siRNA and Negative Control siRNA should be stored at -20°C. |
| Control Type | Negative Control, Positive Control |
| Cell Type | Mammalian |
| Format | Kit |
| Sample Type | Synthetic siRNA, miRNA |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | siRNA Transfection |
| Product Line | Silencer, Ambion |
| Product Type | siRNA Transfection Optimization Kit |
| Quantity | 1 mL kit |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.