Learn More
Microbiologics™ KWIK-STIK™ Staphylococcus epidermidis derived from ATCC™ 35984™†
SDP
Catalog No. 23017270 Shop All Microbiologics Products
Change view
Click to view available options
ATCC®:
ATCC™ 35984™
Catalog No. ATCC®
23-017-270 ATCC™ 35984™
1 options
This item is not returnable. View return policy
Catalog No. 23-017-270 Supplier Microbiologics™ Supplier No. 01189P
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
For microbial identification
Specifications
ATCC® ATCC™ 35984™
Biosafety Level 1
Description KWIK-STIK Staphylococcus epidermidis derived from ATCC™ 35984™†
Detects Staphylococcus epidermidis derived from ATCC™ 35984™
For Use With (Application) Microbial Identification
Incubation Time 24 to 48 hr.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.