Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Staphylococcus Intermedius Z112, Titered
SDP

Catalog No. 21100500 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-100-500 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains live and infectious organisms.
Catalog No. 21-100-500 Supplier ZeptoMetrix Supplier No. 0801592
Add to Cart
Add to Cart
Contains live and infectious organisms.

Used for determination of limit of detection (LOD), in diagnostic assays or crossreactivity studies

  • Biosafety level 2 microorganism
  • Available in 1mL aliquot
  • 16S sequencing used to confirm identification of organism
  • Gram staining with additional culturing monitors purity of culture
  • Glycerol (15%) used as cryoprotectant for freezing medium
  • Also used for nucleic acid tests

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Content And Storage -65°C
Form Solution
Product Type Titered Live Bacteria Control
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.