Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Staphylococcus Lugdunensis Z097, Titered
SDP

Catalog No. 21020102 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-020-102 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains live organism and is potentially biohazardous.
Catalog No. 21-020-102 Supplier ZeptoMetrix Supplier No. 0801555
Add to Cart
Add to Cart
Contains live organism and is potentially biohazardous.

Staphylococcus lugdunensis (coagulase negative Staphylococcus species) consists of gram-positive bacteria with spherical cells that appear in clusters

  • Biosafety level 2 microorganism
  • Available in 1mL frozen aliquot
  • 16S sequencing used to confirm identification of organism
  • Gram staining with additional culturing for monitoring of purity of culture
  • Glycerol (15%) used as cryoprotectant for freezing medium
  • Also used as control for nucleic acid tests

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA prior to shipment.

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Product Type Titered Live Bacteria Control
For Use With (Application) In determination of limit of detection, diagnostic assay development or cross-reactivity studies and as controls in nucleic acid tests
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.