Learn More
Medchemexpress LLC HY-P7228 10ug Medchemexpress, I-TAC/CXCL11 (HEK293-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 502083130
Encompass_Preferred
Medchemexpress, HY-P7228 10ug I-TAC/CXCL11 (HEK293-expressed), Human CAS: Recombinant Human I-TAC/CXCL11 (HEK293-expressed) is a small cytokine belonging to the CXC chemokine family which can be strongly induced by IFN-gamma, and its receptor CXCR3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3130 Supplier Medchemexpress LLC Supplier No. HYP722810UG
Medchemexpress, HY-P7228 10ug I-TAC/CXCL11 (HEK293-expressed), Human CAS: Recombinant Human I-TAC/CXCL11 (HEK293-expressed) is a small cytokine belonging to the CXC chemokine family which can be strongly induced by IFN-gamma, and its receptor CXCR3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.