Learn More
Medchemexpress LLC HY-P7229 50ug Medchemexpress, I-TAC/CXCL11, Human CAS: Purity:>98%
SDP
Catalog No. 501934366
Encompass_Preferred
Medchemexpress, HY-P7229 50ug I-TAC/CXCL11, Human CAS: Recombinant Human I-TAC/CXCL11 is a small cytokine belonging to the CXC chemokine family which can be strongly induced by IFN-gamma, and its receptor CXCR3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4366 Supplier Medchemexpress LLC Supplier No. HYP722950UG
Medchemexpress, HY-P7229 50ug I-TAC/CXCL11, Human CAS: Recombinant Human I-TAC/CXCL11 is a small cytokine belonging to the CXC chemokine family which can be strongly induced by IFN-gamma, and its receptor CXCR3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.