Learn More
Bound Tree Medical Blood/Solution Set, Y-Type, 10 Drops/mL, Interlink Injection Site, each
Catalog No. NC1418353
Encompass_Preferred
Y-type blood/solution set with standard blood filter (170u to 260u filter). Interlink injection site and male luer lock adapter with retractable collar. Contains DEHP. Fluid path is sterile, nonpyrogenic.
This item is not returnable. View return policy
Catalog No. NC1418353 Supplier Bound Tree Medical Supplier No. 1182C6750HEA
Y-type blood/solution set with standard blood filter (170u to 260u filter). Interlink injection site and male luer lock adapter with retractable collar. Contains DEHP. Fluid path is sterile, nonpyrogenic.