Learn More
Phoenix Pharmaceuticals Inc PACAP38 Radioligand, I-125 Isotope, High Affinity for PAC1 Receptor
Catalog No. NC3356357
Encompass
I-125 Labeled PACAP38 is a specialized radioligand designed for precise receptor binding assays. It enables detailed study of PACAP38 interactions with PAC1 and other VPAC receptors, supporting research in neuroprotection, neurodevelopment, nociception, and inflammation. This high-affinity product (Kd = 100 pM) offers rapid and saturable membrane binding, making it ideal for investigating diverse physiological functions and potential therapeutic implications in conditions like migraines.
Catalog No. NC3356357 Supplier Phoenix Pharmaceuticals Inc Supplier No. T05205
I-125 Labeled PACAP38 is a specialized radioligand designed for precise receptor binding assays. It enables detailed study of PACAP38 interactions with PAC1 and other VPAC receptors, supporting research in neuroprotection, neurodevelopment, nociception, and inflammation. This high-affinity product (Kd = 100 pM) offers rapid and saturable membrane binding, making it ideal for investigating diverse physiological functions and potential therapeutic implications in conditions like migraines.