Learn More
Medchemexpress LLC I2959 | 106797-53-9 | 98.7% | 224.26 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW013508
Encompass_Preferred
I2959 is widely used as a photoinitiator for photochemical cross-linking of hydrogels, and the combination of I2959 and CMA hydrogel has been used as a bio-ink for biological 3D printing applications. In addition, I2959 has a high free radical generation efficiency and is cytotoxic to rapidly dividing cell lines.
  • Photoinitiator for photochemical cross-linking of hydrogels
  • Used in combination with CMA hydrogel as a bio-ink for biological 3D printing applications
  • High free radical generation efficiency
  • Cytotoxic to rapidly dividing cell lines
Catalog No. 50-004-53967