Learn More
Waters Corp i2V Assembly, BioACQUITY System, QSM & BSM Compatible
Catalog No. 502391938 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement i2V assembly designed for use with BioACQUITY systems. It is compatible with H-Class Bio Quaternary Solvent Managers and M-Class Binary and Auxiliary Solvent Managers.
  • Replacement i2V assembly
  • Compatible with H-Class Bio Quaternary Solvent Managers
  • Compatible with M-Class Binary and Auxiliary Solvent Managers
Catalog No. 50-239-1938 Supplier Waters Corp Supplier No. 700005412
This is a replacement i2V assembly designed for use with BioACQUITY systems. It is compatible with H-Class Bio Quaternary Solvent Managers and M-Class Binary and Auxiliary Solvent Managers.
  • Replacement i2V assembly
  • Compatible with H-Class Bio Quaternary Solvent Managers
  • Compatible with M-Class Binary and Auxiliary Solvent Managers