Learn More
Waters Corp i2V Cartridge Check Valve, THF/Hexane, UPLC System, Stainless Steel
Catalog No. 502392992 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This i2V Cartridge Check Valve is designed with enhanced compatibility for normal phase solvents. It serves as a replacement component for various systems.
  • Constructed from materials optimized for normal phase solvents.
  • Standard component for APC p-ISM, p-QSM, I-Class, and I-Class PLUS Binary Solvent Managers.
  • Optional component for H-Class and UPLC systems.
  • Supplied as a 2-pack.
  • Made from stainless steel.
  • Designed for UPLC system type.
  • Available as performance maintenance and rebuild kits.
  • Recommended annual replacement schedule.
Catalog No. 50-239-2992 Supplier Waters Corp Supplier No. 700004139
This i2V Cartridge Check Valve is designed with enhanced compatibility for normal phase solvents. It serves as a replacement component for various systems.
  • Constructed from materials optimized for normal phase solvents.
  • Standard component for APC p-ISM, p-QSM, I-Class, and I-Class PLUS Binary Solvent Managers.
  • Optional component for H-Class and UPLC systems.
  • Supplied as a 2-pack.
  • Made from stainless steel.
  • Designed for UPLC system type.
  • Available as performance maintenance and rebuild kits.
  • Recommended annual replacement schedule.