Learn More
Waters Corp Cartridge, I2V, Hexane/THF, UPLC System
Catalog No. 502392094 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The I2V Cartridge, Hexane/THF is designed for use with ACQUITY UPLC systems, including APC, H-Class, I-Class, and Classic models. This cartridge serves as an upgrade kit for various ACQUITY UPLC components.
  • Upgrade kit for UPLC systems
  • Designed for Hexane/THF applications
  • Features a check valve type
  • Compatible with UPLC system types
  • Suitable for H-Class System and Arc instruments
  • Integrates with ISM and QSM components
  • One unit per package
Catalog No. 50-239-2094 Supplier Waters Corp Supplier No. 700005272
The I2V Cartridge, Hexane/THF is designed for use with ACQUITY UPLC systems, including APC, H-Class, I-Class, and Classic models. This cartridge serves as an upgrade kit for various ACQUITY UPLC components.
  • Upgrade kit for UPLC systems
  • Designed for Hexane/THF applications
  • Features a check valve type
  • Compatible with UPLC system types
  • Suitable for H-Class System and Arc instruments
  • Integrates with ISM and QSM components
  • One unit per package