Learn More
Waters Corp i2V Check Valve Cartridge Assembly, Bio/THF
Catalog No. 502392697 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This i2V Check Valve Cartridge Assembly is a replacement part designed for specific ACQUITY H-Class Bio and ACQUITY Premier systems. It is intended for use in systems converted to run normal phase solvents.
  • Replacement part for ACQUITY H-Class Bio and ACQUITY Premier systems.
  • Used in systems converted to run normal phase solvents.
  • Annual replacement schedule.
  • One unit per package.
Catalog No. 50-239-2697 Supplier Waters Corp Supplier No. 700010387
This i2V Check Valve Cartridge Assembly is a replacement part designed for specific ACQUITY H-Class Bio and ACQUITY Premier systems. It is intended for use in systems converted to run normal phase solvents.
  • Replacement part for ACQUITY H-Class Bio and ACQUITY Premier systems.
  • Used in systems converted to run normal phase solvents.
  • Annual replacement schedule.
  • One unit per package.