Learn More
Medchemexpress LLC HY-128206 5mg Medchemexpress, I3MT-3 CAS:459420-09-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1282065MG
Encompass_Preferred
Medchemexpress, HY-128206 5mg I3MT-3 CAS:459420-09-8 I3MT-3 (HMPSNE) is a potent, selective, and cell-membrane permeable inhibitor of 3-Mercaptopyruvate sulfurtransferase (3MST) (IC50=2.7 μM). I3MT-3 is inactive for other H2S/sulfane sulfur-producing enzymes. I3MT-3 targets a persulfurated cysteine residue located in the active site of 3MST. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1157