Learn More
Med Vet International i4 Microscope with BioVID 4K and 13.3" Monitor, J0334QTBV, Each
Catalog No. 5000585463
Encompass_Preferred
Diagnostic-Ready Imaging BioVID 4K 8MP HD Microscope Camera. The BioVID 4K Ultra HD camera from LW Scientific provides stunning resolution and color for client education and live presentations. Mount the camera to any trinocular C-mount, and you will have access to high quality, 8 megapixel imaging and 4K video on your monitor or television screen. Save images and videos to the camera's SD card, or connect the camera to your computer via USB. The BioVID 4K also has an HDMI port for connection to any large HDTV or projector. With a resolution of 3840 x 2160, the BioVID 4K camera will display 4K quality onto any 4K compatible screen. This camera comes equipped with an extremely sensitive Sony IMX334(C) sensor chip to help it maintain 4K resolution and a fast frame rate in low-light and high magnification settings. No matter where you are, the BioVID 4K will ensure a crisp, clear image from the lecture hall to the laboratory. Connections: USB HDMI Performance: 8 mp @ 30 fps
Catalog No. 50-005-85463 Supplier Med Vet International Supplier No. JOR334QTBV
Diagnostic-Ready Imaging BioVID 4K 8MP HD Microscope Camera. The BioVID 4K Ultra HD camera from LW Scientific provides stunning resolution and color for client education and live presentations. Mount the camera to any trinocular C-mount, and you will have access to high quality, 8 megapixel imaging and 4K video on your monitor or television screen. Save images and videos to the camera's SD card, or connect the camera to your computer via USB. The BioVID 4K also has an HDMI port for connection to any large HDTV or projector. With a resolution of 3840 x 2160, the BioVID 4K camera will display 4K quality onto any 4K compatible screen. This camera comes equipped with an extremely sensitive Sony IMX334(C) sensor chip to help it maintain 4K resolution and a fast frame rate in low-light and high magnification settings. No matter where you are, the BioVID 4K will ensure a crisp, clear image from the lecture hall to the laboratory. Connections: USB HDMI Performance: 8 mp @ 30 fps