Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ i5100 Autodetect Label Roll Holder

Catalog No. 19129134 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-129-134 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-134 Supplier Brady™ Supplier No. 149470
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brady™ i5100 Autodetect Label Roll Holder enables materials for faster and easier material changeovers and set up

  • For use with the BradyPrinter i5100 Industrial Label Printer (also fits in Brady IP Printer)
  • Use with IP-enabled label rolls for easier faster setup including auto sensor-select, on-screen part information and auto label setup in Brady software
  • Holds 3in. open core media rolls

Specifications

For Use With (Equipment) BradyPrinter i5100 Industrial Label Printer
Width (English) 6.6 in.
Height (English) 5.7 in.
Depth (English) 4.25 in.
Description Media Handling Accessories
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.