Learn More
Brady™ i7500 Industrial Label Printer Perforation Cutter Accessory Black Each
Catalog No. 19221512 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
i7500
Catalog No. For Use With (Equipment)
19-221-512 i7500
1 options
Catalog No. 19-221-512 Supplier Brady™ Supplier No. 177725
Add to Cart
Add to Cart
For use with the Standard model only.
  • Enables partial-perf or full cut through of approved printed materials
  • Can be set to various cut depths, full cut through or combinations of both in the same print job
  • Cut location can be set to cut after print job, after every label row, after label set or to not cut at all
  • Ideal for use on continuous Brady PermaSleeve material
  • Cut results vary depending on material and length of cut label; user should test on intended material and application
  • Installs easily with four screws using included T-20 wrench that ships with all i7500 printer models. Auto Cutter accessories are not compatible with i7500 Peel models
Specifications
Product Type Cutter Tool
For Use With (Equipment) i7500