Learn More
Description
- Get 40-second part changeovers with no adjustments, no trial-and-error print tests and no wasted labels. LabelSense™ Technology allows the printer, Brady i75-Series materials and Brady software to communicate; automating and eliminating tedious adjustments and label-wasting tasks.
Specifications
Specifications
| Product Type | Label Printer |
| Color | Blue, Gray |
| Display Type | Color LCD Touchscreen |
| For Use With (Equipment) | 2HX Series Printers, HX Series Printers, 3PS Series Printers, THT Series Printers, Brady Workstation |
| Material | Plastic |
| Print Technology | Direct Thermal, Thermal Transfer |
| Width (English) | 11.5 in |
| Resolution | 300 dpi |
| Software Type | Product and Wire ID Software |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.