Learn More
Brady™ i7500 Printer Accessory 1.5 in. Internal Rewind Takeup Core, Brown
Catalog No. 18038073 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
i7500
Catalog No. For Use With (Equipment)
18-038-073 i7500
1 options
Catalog No. 18-038-073 Supplier Brady™ Supplier No. 177727
Add to Cart
Add to Cart
For use with the i7500 Industrial Label Printer - Peel model only.
  • Replacement or spare core (Peel configuration printer ships with qty 1 of this empty core). Required when using peel configuration printer to internally rewind printed labels. 1.5 in. (38.1 mm) inside diameter corrugated core that fits onto internal takeup spindle of Peel configuration printer. Internally-rewound printed labels collect onto this core for easy removal and easier roll handling after done printing.
Specifications
Product Type Label Tray
For Use With (Equipment) i7500