Learn More
Brady™ i7500 Printer Accessory Peel Plate
Catalog No. 18038076 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
i7500
Catalog No. For Use With (Equipment)
18-038-076 i7500
1 options
Catalog No. 18-038-076 Supplier Brady™ Supplier No. 177731
Add to Cart
Add to Cart
For use with the i7500 Industrial Label Printer - Peel model only.
  • Replacement plate (Peel configuration printer ships with this plate installed). Smooth-edge plate automatically peels qualified auto-dispensale labels from liner in peel mode, or can be used to tear thin paper liners for non-auto-peel printing. Easily removed and installed using the included T-20 wrench that ships with all i7500 printer models. Peel results vary depending on the auto-dispense capability of the material and label size; user should test on intended material and application.
Specifications
Product Type Printer Plate
For Use With (Equipment) i7500