Learn More
Brady™ i7500 Printer Accessory Print Roller
Catalog No. 19030002 Shop All Brady Corporation Products
Change view
Click to view available options
Width (Metric):
25 mm
50 mm
80 mm
114 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19-030-002 25 mm
19-029-999 114 mm
19-030-000 80 mm
19-030-001 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-030-002 Supplier Brady™ Supplier No. 177737
Add to Cart
Add to Cart
For use with the i7500 Industrial Label Printer.
  • Use narrowest roller that covers the media width, and use narrowest ribbon that covers both media and roller.
Specifications
Product Type Alignment Roll
For Use With (Equipment) i7500
Width (Metric) 25 mm