Learn More
Brady i7500 Printer Accessory Replacement 600dpi Printhead
Catalog No. 17100967 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
i7500 Label Printer
Catalog No. For Use With (Equipment)
17-100-967 i7500 Label Printer
1 options
Catalog No. 17-100-967 Supplier Brady Supplier No. 177723
Add to Cart
Add to Cart
i7500 Industrial Label Printer, ideal for high-precision detail printing including smaller fonts, high-density linear barcodes and 2D barcodes
  • Only Brady i7500 print heads can be used in the i7500 printer
  • Printhead replacements or head dpi changes are highly recommended to be completed by Brady Technical Support, which includes ESD-handling and factory fine-tuning with head alignment
  • Printheads can be changed by the user to a replacement 300 dpi head or a 600 dpi head so long as the printer has firmware v2.0 or above (ESD grounding and gloves recommended)
  • To optimize printhead life, clean the head regularly with IPA cleaning swabs (PCK 6), replace worn or rough platen rollers, use narrow rollers when printing narrow ribbon or narrow label combinations and avoid unnecessary high printhead heat settings
  • Damage to the printhead due to user installation is not covered under warranty
  • Warranty on replacement printheads is 1 year
Specifications
Product Type Printhead
For Use With (Equipment) i7500 Label Printer