Learn More
Medchemexpress LLC IA9 TFA (human TREM-2 182-190 TFA) | 98.9% | 1001.31 (free acid) | 5 MG
SDP
Catalog No. 5000424718
Encompass_Preferred
IA9 TFA is a potent, CNS-penetrant inhibitor of Triggering Receptor Expressed on Myeloid cells 2 (TREM-2). It is capable of reducing proinflammatory cytokines and mitigating joint inflammation and damage in a Collagen-induced arthritis mouse model. This compound is suitable for neuroinflammation PET imaging and research related to rheumatoid arthritis and neuroinflammation.
Features and Benefits
  • Potent CNS-penetrant TREM-2 inhibitor.
  • Diminishes the release of proinflammatory cytokines.
  • Suppresses joint inflammation and damage in experimental arthritis.
  • Can be used for neuroinflammation PET imaging.
  • Applicable for rheumatoid arthritis and neuroinflammation research.
Catalog No. 50-004-24718 Supplier Medchemexpress LLC Supplier No. HYP10136A5MG
IA9 TFA is a potent, CNS-penetrant inhibitor of Triggering Receptor Expressed on Myeloid cells 2 (TREM-2). It is capable of reducing proinflammatory cytokines and mitigating joint inflammation and damage in a Collagen-induced arthritis mouse model. This compound is suitable for neuroinflammation PET imaging and research related to rheumatoid arthritis and neuroinflammation.
Features and Benefits
  • Potent CNS-penetrant TREM-2 inhibitor.
  • Diminishes the release of proinflammatory cytokines.
  • Suppresses joint inflammation and damage in experimental arthritis.
  • Can be used for neuroinflammation PET imaging.
  • Applicable for rheumatoid arthritis and neuroinflammation research.