Learn More
Apexbio Technology LLC IAA-94 54197-31-8 10mg
SDP
Catalog No. 5000091573
Encompass_Preferred
IAA-94 (CAS 54197-31-8) is a small-molecule inhibitor of epithelial chloride channels modulating cellular electrophysiological properties by disrupting chloride ion transport Experimental studies have demonstrated that IAA-94 can interfere with interactions involving the negative factor (Nef) protein during infection indicating its capacity to modulate Nef-mediated processes Due to its specificity in targeting chloride channels IAA-94 is widely utilized in investigations of ion channel-related signaling pathways in tumor cells and serves as a valuable tool in studies of cell electrophysiology and infection mechanisms
Catalog No. 50-000-91573 Supplier Apexbio Technology LLC Supplier No. C501010
IAA-94 (CAS 54197-31-8) is a small-molecule inhibitor of epithelial chloride channels modulating cellular electrophysiological properties by disrupting chloride ion transport Experimental studies have demonstrated that IAA-94 can interfere with interactions involving the negative factor (Nef) protein during infection indicating its capacity to modulate Nef-mediated processes Due to its specificity in targeting chloride channels IAA-94 is widely utilized in investigations of ion channel-related signaling pathways in tumor cells and serves as a valuable tool in studies of cell electrophysiology and infection mechanisms