Learn More
Medchemexpress LLC IACS-010759 | 1570496-34-2 | 99.7% | 562.56 | 100 MG
SDP
Catalog No. 5000439356
Encompass_Preferred
IACS-010759 is an orally active, potent mitochondrial complex I inhibitor of oxidative phosphorylation (OXPHOS). It inhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS, making it suitable for research in relapsed/refractory AML and solid tumors.
  • Potent mitochondrial complex I inhibitor
  • Inhibits proliferation and induces apoptosis in cancer models
  • Potential for solid tumor research
  • Suitable for acute myeloid leukemia (AML) research
  • Exhibits low plasma clearance and prolonged terminal half-life
Catalog No. 50-004-39356 Supplier Medchemexpress LLC Supplier No. HY112037A100MG
IACS-010759 is an orally active, potent mitochondrial complex I inhibitor of oxidative phosphorylation (OXPHOS). It inhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS, making it suitable for research in relapsed/refractory AML and solid tumors.
  • Potent mitochondrial complex I inhibitor
  • Inhibits proliferation and induces apoptosis in cancer models
  • Potential for solid tumor research
  • Suitable for acute myeloid leukemia (AML) research
  • Exhibits low plasma clearance and prolonged terminal half-life