Learn More
Medchemexpress LLC HY-112037 5mg , IACS-010759 CAS:1570496-34-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1120375MG
Encompass_Preferred
HY-112037 5mg IACS-010759 CAS: 1570496-34-2 IACS-010759 is an orally active, potent mitochondrial complex I of oxidative phosphorylation (OXPHOS) inhibitor. IACS-010759 inhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS. IACS-010759 has the potential for relapsed/refractory AML and solid tumors research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9227